Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256672.1 | NP_001243601.1 | 2 | 292 | ||
NM_001256673.1 | NP_001243602.1 | 2 | 292 | |||
NM_001256674.1 | NP_001243603.1 | 2 | 292 | |||
NM_001256675.1 | NP_001243604.1 | 2 | 292 | |||
NM_001256676.1 | NP_001243605.1 | 2 | 106 | |||
NM_001256677.1 | NP_001243606.1 | 3 | 423 | |||
NM_001324226.1 | NP_001311155.1 | 2 | 106 | |||
NM_001324227.1 | NP_001311156.1 | 2 | 106 | |||
NM_001324228.1 | NP_001311157.1 | 2 | 170 | |||
NM_001324229.1 | NP_001311158.1 | 2 | 292 | |||
NM_001324230.1 | NP_001311159.1 | 3 | 423 | |||
NM_004809.4 | NP_004800.2 | 2 | 292 | |||
XM_005254792.4 | XP_005254849.1 | 2 | 1196 | |||
XM_006720773.3 | XP_006720836.1 | 2 | 1197 | |||
GenBank mRNA | AF074953.1 | - | 2 | 154 | ||
AF111800.1 | - | 2 | 241 | |||
AF156557.1 | - | 2 | 300 | |||
AK075244.1 | - | 2 | 188 | |||
AK091531.1 | - | 2 | 92 | |||
AK222665.1 | - | 2 | 175 | |||
AK300799.1 | - | 3 | 307 | |||
AK302051.1 | - | 2 | 247 | |||
AL109664.1 | - | 2 | 122 | |||
AL109665.1 | - | 2 | 146 | |||
AL533460.3 | - | 2 | 83 | |||
BC034379.1 | - | 2 | 238 | 5185908 | ||
BC065249.1 | - | 2 | 238 | 6059393 | ||
BX464941.2 | - | 2 | 106 | |||
CR542089.1 | - | 2 | 167 | |||
DQ064605.1 | - | 2 | 167 | |||
Y16522.1 | - | 2 | 211 |