Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033198.3 | NP_149975.1 | 12 | 1868 | ||
GenBank mRNA | AB057723.1 | - | 12 | 1495 | ||
AF161427.1 | - | 6 | 730 | |||
AK025825.1 | - | 5 | 633 | |||
AK055512.1 | - | 7 | 698 | |||
AK075428.1 | - | 12 | 1523 | |||
AK127162.1 | - | 3 | 1436 | |||
AK297046.1 | - | 11 | 1387 | |||
AK314583.1 | - | 12 | 1531 | |||
AY359112.1 | - | 11 | 1763 | |||
BC001319.2 | - | 4 | 561 | |||
BC007301.1 | - | 10 | 1252 | |||
BC069228.1 | - | 12 | 1518 | 3944034 |