Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004814.2 | NP_004805.2 | 4 | 506 | ||
GenBank mRNA | AF090988.1 | - | 4 | 475 | ||
AK074516.1 | - | 4 | 489 | |||
AK298823.1 | - | 4 | 524 | |||
AK313800.1 | - | 4 | 471 | |||
BC001494.2 | - | 4 | 482 | 3504573 | ||
EF011621.1 | - | 4 | 481 |