Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004815.3 | NP_004806.3 | 14 | 1927 | ||
XM_006711048.3 | XP_006711111.1 | 13 | 1561 | |||
XM_011542438.2 | XP_011540740.1 | 14 | 1802 | |||
XM_011542439.2 | XP_011540741.1 | 14 | 1647 | |||
XM_011542440.2 | XP_011540742.1 | 13 | 1521 | |||
GenBank mRNA | AB208873.1 | - | 13 | 1533 | ||
AK307733.1 | - | 14 | 1756 | |||
AK310491.1 | - | 14 | 1647 | |||
AL833445.1 | - | 2 | 6136 | |||
BC093741.1 | - | 13 | 1600 | 7939586 | ||
BC093767.1 | - | 13 | 1603 | 7939612 | ||
U90920.1 | - | 14 | 1741 |