Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TJP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170414.2 | NP_001163885.1 | 5 | 449 | ||
NM_001170415.1 | NP_001163886.1 | 4 | 449 | |||
NM_001170416.1 | NP_001163887.1 | 4 | 379 | |||
NM_004817.3 | NP_004808.2 | 4 | 604 | |||
NM_201629.3 | NP_963923.1 | 4 | 604 | |||
XM_005252314.2 | XP_005252371.1 | 4 | 657 | |||
XM_011519204.1 | XP_011517506.1 | 4 | 347 | |||
XM_011519206.2 | XP_011517508.1 | 6 | 949 | |||
XM_011519207.2 | XP_011517509.1 | 5 | 445 | |||
XM_011519208.2 | XP_011517510.1 | 5 | 405 | |||
XM_011519209.2 | XP_011517511.1 | 3 | 234 | |||
XM_017015326.1 | XP_016870815.1 | 5 | 502 | |||
XM_017015327.1 | XP_016870816.1 | 4 | 604 | |||
XM_017015328.1 | XP_016870817.1 | 4 | 657 | |||
GenBank mRNA | AF083892.1 | - | 4 | 415 | ||
AF083893.1 | - | 5 | 603 | |||
AF489824.1 | - | 4 | 365 | |||
AI738923.1 | - | 4 | 340 | |||
AK295016.1 | - | 5 | 405 | |||
AK295034.1 | - | 4 | 379 | |||
AK302483.1 | - | 4 | 305 | |||
AK304447.1 | - | 6 | 531 | |||
BC027592.1 | - | 4 | 495 | 4839233 | ||
DC325189.1 | - | 4 | 449 | |||
L27476.1 | - | 4 | 365 |