Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281501.1 | NP_001268430.1 | 4 | 619 | ||
NM_001281502.1 | NP_001268431.1 | 4 | 688 | |||
NM_004265.3 | NP_004256.1 | 4 | 960 | |||
GenBank mRNA | AF084559.1 | - | 4 | 752 | ||
AF126799.1 | - | 4 | 615 | |||
AK074939.1 | - | 4 | 692 | |||
AK074991.1 | - | 4 | 428 | |||
AK290291.1 | - | 4 | 566 | |||
AK299762.1 | - | 4 | 688 | |||
AL050118.1 | - | 2 | 204 | |||
BC009011.1 | - | 4 | 694 | 4181037 |