Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143970.1 | NP_001137442.1 | 5 | 1379 | ||
NM_001143971.1 | NP_001137443.1 | 5 | 1303 | |||
NM_004824.3 | NP_004815.3 | 5 | 1530 | |||
NR_026590.1 | 7 | 1485 | ||||
XM_011514997.2 | XP_011513299.1 | 5 | 1459 | |||
GenBank mRNA | AF081258.1 | - | 7 | 1485 | ||
AF081259.1 | - | 7 | 1485 | |||
AK291601.1 | - | 6 | 1293 | |||
AK296985.1 | - | 5 | 1379 | |||
BC061516.1 | - | 4 | 600 | |||
BC108725.1 | - | 5 | 1541 | 6140263 | ||
BC119682.2 | - | 4 | 526 | 40116323 |