Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244.3 | NP_001235.1 | 1 | 466 | ||
NM_001252290.1 | NP_001239219.1 | 1 | 466 | |||
GenBank mRNA | AK313619.1 | - | 1 | 466 | ||
AL583443.3 | - | 1 | 370 | |||
BC093630.1 | - | 1 | 248 | 7939475 | ||
BC111939.1 | - | 1 | 248 | 8327407 | ||
CD690231.1 | - | 1 | 176 | |||
L09753.1 | - | 1 | 288 |