Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244.3 | NP_001235.1 | 4 | 702 | ||
GenBank mRNA | AK313619.1 | - | 4 | 702 | ||
AL583443.3 | - | 4 | 606 | |||
BC093630.1 | - | 4 | 484 | 7939475 | ||
BC111939.1 | - | 4 | 484 | 8327407 | ||
L09753.1 | - | 4 | 524 |