Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033677.1 | NP_001028849.1 | 3 | 774 | ||
NM_004276.4 | NP_004267.2 | 3 | 229 | |||
NM_031205.3 | NP_112482.1 | 4 | 409 | |||
XM_017020235.1 | XP_016875724.1 | 3 | 841 | |||
XM_017020236.1 | XP_016875725.1 | 3 | 976 | |||
XM_017020237.1 | XP_016875726.1 | 3 | 976 | |||
XM_017020238.1 | XP_016875727.1 | 4 | 2055 | |||
XM_017020239.1 | XP_016875728.1 | 3 | 1869 | |||
GenBank mRNA | AF169148.1 | - | 3 | 168 | ||
AF169149.1 | - | 4 | 348 | |||
AK127203.1 | - | 1 | 1243 | |||
AL533429.3 | - | 3 | 743 | |||
BC030201.1 | - | 3 | 774 | 5200625 | ||
BM716422.1 | - | 3 | 229 |