Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204051.1 | NP_001190980.1 | 2, 3 | 566 | ||
NM_001204052.1 | NP_001190981.1 | 2, 3 | 566 | |||
NM_004277.4 | NP_004268.3 | 2, 3 | 566 | |||
XM_005249483.2 | XP_005249540.1 | 2, 3 | 536 | |||
GenBank mRNA | AF110532.1 | - | 2, 3 | 288 | ||
AK291427.1 | - | 2, 3 | 496 | |||
AK295233.1 | - | 2, 3 | 549 | |||
AK315940.1 | - | 2, 3 | 477 | |||
AY358711.1 | - | 2, 3 | 314 | |||
BC033091.1 | - | 2, 3 | 539 | 5455903 | ||
BP310559.1 | - | 3, 4 | 453 | |||
DA144842.1 | - | 540 | ||||
DR002070.1 | - | 2, 3 | 488 |