Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMPCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004279.2 | NP_004270.2 | 9 | 1059 | ||
XM_005250717.3 | XP_005250774.1 | 9 | 1073 | |||
XM_006716181.3 | XP_006716244.1 | 9 | 1072 | |||
GenBank mRNA | AF054182.1 | - | 9 | 1038 | ||
AK000994.1 | - | 9 | 1041 | |||
AK057656.1 | - | 8 | 857 | |||
AK098616.1 | - | 9 | 1029 | |||
AK289864.1 | - | 9 | 1041 | |||
AK297354.1 | - | 9 | 1424 | |||
AL050216.1 | - | 5 | 508 | |||
BC014079.2 | - | 9 | 999 |