Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004863.3 | NP_004854.1 | 4 | 686 | ||
XM_011537384.2 | XP_011535686.1 | 4 | 582 | |||
GenBank mRNA | AB011098.1 | - | 4 | 686 | ||
BC005123.2 | - | 4 | 587 | 3688136 | ||
U15555.1 | - | 3 | 322 | |||
Y08686.1 | - | 4 | 546 |