Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170714.1 | NP_001164185.1 | 5 | 1269 | ||
NM_001170715.1 | NP_001164186.1 | 4 | 974 | |||
NM_001170716.1 | NP_001164187.1 | 4 | 931 | |||
NM_001170717.1 | NP_001164188.1 | 4 | 993 | |||
NM_001170718.1 | NP_001164189.1 | 4 | 1018 | |||
NM_001170719.1 | NP_001164190.1 | 4 | 1108 | |||
NM_001170720.1 | NP_001164191.1 | 5 | 679 | |||
NM_001170721.1 | NP_001164192.1 | 3 | 1206 | |||
NM_014567.3 | NP_055382.2 | 4 | 993 | |||
XM_005256258.4 | XP_005256315.1 | 4 | 1193 | |||
XM_005256259.4 | XP_005256316.1 | 4 | 4105 | |||
XM_005256260.4 | XP_005256317.1 | 4 | 1114 | |||
XM_011523475.2 | XP_011521777.1 | 4 | 1142 | |||
XM_017023897.1 | XP_016879386.1 | 4 | 1700 | |||
XM_017023898.1 | XP_016879387.1 | 4 | 1700 | |||
GenBank mRNA | AB040024.1 | - | 4 | 988 | ||
AF218451.1 | - | 4 | 971 | |||
AJ242987.2 | - | 4 | 968 | |||
AK027608.1 | - | 4 | 1018 | |||
AK074958.1 | - | 3 | 403 | |||
AK124526.1 | - | 4 | 2413 | |||
AK124815.1 | - | 4 | 977 | |||
AK124844.1 | - | 3 | 784 | |||
AK293808.1 | - | 5 | 1269 | |||
AK294513.1 | - | 4 | 1024 | |||
AK295809.1 | - | 4 | 903 | |||
AK302617.1 | - | 5 | 679 | |||
AK314722.1 | - | 4 | 993 | |||
AY545071.1 | - | 3 | 847 | |||
BC062556.1 | - | 3 | 848 | 6427808 |