Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042385.2 | NP_001035844.1 | 11 | 1803 | ||
NM_001042386.2 | NP_001035845.1 | 11 | 1791 | |||
NM_001171603.1 | NP_001165074.1 | 13 | 2329 | |||
NM_001171606.1 | NP_001165077.1 | 13 | 2460 | |||
NM_001171613.1 | NP_001165084.1 | 12 | 1918 | |||
NM_001171617.1 | NP_001165088.1 | 12 | 2075 | |||
NM_006036.4 | NP_006027.2 | 12 | 2914 | |||
XM_011533198.1 | XP_011531500.1 | 13 | 2416 | |||
XM_011533200.1 | XP_011531502.1 | 13 | 2068 | |||
XM_011533202.1 | XP_011531504.1 | 12 | 1868 | |||
XM_017005384.1 | XP_016860873.1 | 13 | 3054 | |||
XM_017005385.1 | XP_016860874.1 | 13 | 2379 | |||
GenBank mRNA | AB007896.1 | - | 12 | 1841 | ||
AK025649.1 | - | 12 | 1761 | |||
AK131463.1 | - | 13 | 2272 | |||
BC013193.1 | - | 4 | 506 | |||
BC151236.1 | - | 13 | 2393 | 8860445 | ||
DQ023503.1 | - | 12 | 2914 | |||
DQ023504.1 | - | 12 | 2914 | |||
DQ023505.1 | - | 12 | 2914 | |||
DQ023506.1 | - | 11 | 1803 | |||
DQ023507.1 | - | 11 | 1791 |