Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284259.1 | NP_001271188.1 | 23, 24 | 4187 | ||
NM_016195.3 | NP_057279.2 | 23, 24 | 4067 | |||
GenBank mRNA | AB033337.1 | - | 23, 24 | 4140 | ||
AK024959.1 | - | 6, 7 | 1762 | |||
AK025628.1 | - | 3, 4 | 222 | |||
AL117496.1 | - | 23, 24 | 4022 | |||
AL137392.1 | - | 10, 11 | 2654 | |||
AY282407.1 | - | 8, 9 | 1984 | |||
AY739715.1 | - | 4, 5 | 780 | |||
L16782.1 | - | 4, 5 | 334 | |||
U93121.1 | - | 4, 5 | 895 |