Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000666.2 | NP_000657.1 | 4 | 500 | ||
NM_001198895.1 | NP_001185824.1 | 4 | 495 | |||
NM_001198896.1 | NP_001185825.1 | 4 | 500 | |||
NM_001198897.1 | NP_001185826.1 | 4 | 500 | |||
GenBank mRNA | AK298274.1 | - | 4 | 249 | ||
BC000545.2 | - | 4 | 244 | 3162222 | ||
BC003023.1 | - | 4 | 237 | 2821665 | ||
BC014112.2 | - | 4 | 220 | 4543442 | ||
CD014023.1 | - | 4 | 341 | |||
CD014026.1 | - | 4 | 341 | |||
D14524.1 | - | 4 | 234 | |||
D16307.1 | - | 4 | 232 | |||
EU794586.1 | - | 4 | 220 | |||
KU177890.1 | - | 3 | 176 | |||
L07548.1 | - | 4 | 237 |
Gene Symbol: ABHD14A-ACY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316331.1 | NP_001303260.1 | 6 | 701 | ||
GenBank mRNA | AK294015.1 | - | 3 | 274 | ||
AK298086.1 | - | 6 | 677 |