Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004913.2 | NP_004904.2 | 6 | 695 | ||
XM_005256329.4 | XP_005256386.1 | 6 | 755 | |||
XM_005256330.4 | XP_005256387.1 | 6 | 572 | |||
XM_005256331.2 | XP_005256388.1 | 3 | 229 | |||
XM_006721350.2 | XP_006721413.1 | 6 | 755 | |||
XM_011523476.2 | XP_011521778.1 | 6 | 752 | |||
XM_011523477.2 | XP_011521779.1 | 5 | 2568 | |||
XM_011523478.2 | XP_011521780.1 | 5 | 2565 | |||
XM_011523479.2 | XP_011521781.1 | 6 | 755 | |||
XM_011523480.2 | XP_011521782.1 | 6 | 609 | |||
GenBank mRNA | AB018551.1 | - | 5 | 736 | ||
BY795348.2 | - | 3 | 316 |