Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014630.2 | NP_055445.2 | 4 | 1991 | ||
XM_005254996.3 | XP_005255053.1 | 3 | 1892 | |||
XM_011522246.2 | XP_011520548.1 | 4 | 2061 | |||
XM_011522247.2 | XP_011520549.1 | 3 | 2064 | |||
XM_011522248.2 | XP_011520550.1 | 4 | 2022 | |||
XM_017022734.1 | XP_016878223.1 | 2 | 1772 | |||
GenBank mRNA | BC094688.1 | - | 4 | 1943 | 30397050 | |
BC112232.1 | - | 2 | 1725 | 8327700 | ||
BC112234.1 | - | 2 | 1726 | 8327702 | ||
BC143600.1 | - | 2 | 1727 | |||
D86966.1 | - | 4 | 2225 |