Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014643.3 | NP_055458.1 | 4 | 2301 | ||
XM_011526269.1 | XP_011524571.1 | 3 | 2239 | |||
XM_011526270.2 | XP_011524572.1 | 3 | 2502 | |||
XM_011526271.2 | XP_011524573.1 | 4 | 2616 | |||
XM_011526272.2 | XP_011524574.1 | 4 | 2337 | |||
XM_011526273.2 | XP_011524575.1 | 3 | 2252 | |||
XM_011526274.2 | XP_011524576.1 | 3 | 2258 | |||
XM_011526275.2 | XP_011524577.1 | 3 | 3115 | |||
XM_011526276.1 | XP_011524578.1 | 3 | 2239 | |||
XM_011526277.2 | XP_011524579.1 | 3 | 2240 | |||
XM_017026097.1 | XP_016881586.1 | 4 | 2372 | |||
GenBank mRNA | BC108287.1 | - | 1 | 155 | ||
D86975.1 | - | 4 | 2301 |