Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC653513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003021.1 | XP_016858510.1 | 4 | 815 | ||
XM_017003022.1 | XP_016858511.1 | 4 | 815 | |||
XM_017003024.1 | XP_016858513.1 | 4 | 815 | |||
XM_017003025.1 | XP_016858514.1 | 4 | 815 | |||
XM_017003026.1 | XP_016858515.1 | 4 | 815 | |||
XM_017003027.1 | XP_016858516.1 | 4 | 650 | |||
XM_017003031.1 | XP_016858520.1 | 4 | 650 | |||
XM_017003032.1 | XP_016858521.1 | 4 | 650 | |||
XM_017003033.1 | XP_016858522.1 | 4 | 650 | |||
XM_017003039.1 | XP_016858528.1 | 4 | 815 | |||
XM_017003040.1 | XP_016858529.1 | 2 | 158 |