Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000611.5 | NP_000602.1 | 2, 3 | 193 | ||
NM_001127223.1 | NP_001120695.1 | 1, 2 | 359 | |||
NM_001127225.1 | NP_001120697.1 | 2, 3 | 252 | |||
NM_001127226.1 | NP_001120698.1 | 2, 3 | 187 | |||
NM_001127227.1 | NP_001120699.1 | 2, 3 | 131 | |||
NM_203329.2 | NP_976074.1 | 3, 4 | 309 | |||
NM_203330.2 | NP_976075.1 | 4, 5 | 354 | |||
NM_203331.2 | NP_976076.1 | 3, 4 | 238 | |||
GenBank mRNA | AK128571.1 | - | 1 | 2150 | ||
AK309894.1 | - | 1, 2 | 359 | |||
AK311778.1 | - | 2, 3 | 136 | |||
AL037045.3 | - | 2, 3 | 166 | |||
AU136899.1 | - | 3, 4 | 238 | |||
AU139009.1 | - | 2, 3 | 131 | |||
AU139541.1 | - | 2, 3 | 187 | |||
BC001506.2 | - | 2, 3 | 117 | 2988140 | ||
BI752924.1 | - | 4, 5 | 289 | |||
BQ230563.1 | - | 3, 4 | 157 | |||
BT007104.1 | - | 1, 2 | 77 | |||
CR407661.1 | - | 1, 2 | 77 | |||
CR541722.1 | - | 1, 2 | 77 | |||
DA250543.1 | - | 4, 5 | 300 | |||
DA578905.1 | - | 3, 4 | 253 | |||
DA840870.1 | - | 2, 3 | 253 | |||
DA917093.1 | - | 3, 4 | 220 | |||
M27909.1 | - | 2, 3 | 125 | |||
M34671.1 | - | 1, 2 | 106 | |||
M95708.1 | - | 2, 3 | 139 | |||
X16447.1 | - | 2, 3 | 140 | |||
X17198.1 | - | 2, 3 | 149 |