Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040059.1 | NP_001035148.1 | 4 | 813 | ||
NM_001251.2 | NP_001242.2 | 4 | 895 | |||
GenBank mRNA | AK222492.1 | - | 4 | 742 | ||
AK222514.1 | - | 4 | 741 | |||
AK290238.1 | - | 4 | 755 | |||
AK298055.1 | - | 5 | 702 | |||
AK301223.1 | - | 4 | 647 | |||
BC015557.2 | - | 4 | 767 | 3050731 | ||
BT009923.1 | - | 4 | 704 | |||
BX438711.2 | - | 4 | 652 | |||
BX647524.1 | - | 4 | 662 | |||
S57235.1 | - | 4 | 719 |