Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037324.2 | NP_001032401.1 | 11 | 1455 | ||
NM_001100120.1 | NP_001093590.1 | 12 | 1680 | |||
NM_001100121.1 | NP_001093591.1 | 12 | 1542 | |||
NM_014693.3 | NP_055508.3 | 12 | 1791 | |||
GenBank mRNA | AB011176.1 | - | 12 | 1537 | ||
AF192531.1 | - | 12 | 1447 | |||
AF428263.1 | - | 12 | 1719 | |||
AF428264.1 | - | 11 | 1360 | |||
AF521189.1 | - | 12 | 1656 | |||
AK293891.1 | - | 11 | 1385 | |||
AK295270.1 | - | 5 | 592 | |||
AK296541.1 | - | 11 | 1438 | |||
AK309236.1 | - | 9 | 1169 | |||
AY359003.1 | - | 11 | 1411 | |||
BC142950.1 | - | 11 | 1455 | 8860334 |