Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037324.2 | NP_001032401.1 | 12, 13 | 1600 | ||
NM_001100120.1 | NP_001093590.1 | 13, 14 | 1825 | |||
NM_001100121.1 | NP_001093591.1 | 13, 14 | 1687 | |||
NM_014693.3 | NP_055508.3 | 13, 14 | 1936 | |||
GenBank mRNA | AB011176.1 | - | 13, 14 | 1682 | ||
AF192531.1 | - | 13, 14 | 1592 | |||
AF428263.1 | - | 13, 14 | 1864 | |||
AF428264.1 | - | 12, 13 | 1505 | |||
AF521189.1 | - | 13, 14 | 1801 | |||
AK293891.1 | - | 12, 13 | 1530 | |||
AK295270.1 | - | 6, 7 | 737 | |||
AK296541.1 | - | 12, 13 | 1583 | |||
AK309236.1 | - | 10, 11 | 1314 | |||
AY359003.1 | - | 12, 13 | 1556 | |||
BC142950.1 | - | 12, 13 | 1600 | 8860334 |