Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014696.3 | NP_055511.2 | 3 | 836 | ||
XM_005277666.3 | XP_005277723.1 | 4 | 1112 | |||
XM_011540388.2 | XP_011538690.1 | 6 | 1472 | |||
XM_011540395.2 | XP_011538697.1 | 4 | 1080 | |||
XM_011540400.2 | XP_011538702.1 | 3 | 957 | |||
XM_017016974.1 | XP_016872463.1 | 6 | 3377 | |||
XM_017016975.1 | XP_016872464.1 | 5 | 3711 | |||
XM_017016976.1 | XP_016872465.1 | 6 | 3652 | |||
XM_017016977.1 | XP_016872466.1 | 6 | 1884 | |||
XM_017016978.1 | XP_016872467.1 | 5 | 3765 | |||
XM_017016979.1 | XP_016872468.1 | 4 | 1645 | |||
XM_017016980.1 | XP_016872469.1 | 4 | 1607 | |||
XM_017016981.1 | XP_016872470.1 | 5 | 3222 | |||
XM_017016982.1 | XP_016872471.1 | 5 | 3238 | |||
XM_017016983.1 | XP_016872472.1 | 5 | 3513 | |||
XM_017016984.1 | XP_016872473.1 | 5 | 3497 | |||
GenBank mRNA | AB011086.1 | - | 1 | 1652 | ||
BC011672.2 | - | 3 | 836 | 4300443 |