Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014713.4 | NP_055528.1 | 2 | 705 | ||
GenBank mRNA | AK129875.1 | - | 2 | 355 | ||
AY359028.1 | - | 2 | 321 | |||
BC000421.2 | - | 2 | 282 | 2964450 | ||
BC003158.1 | - | 2 | 345 | 2958870 | ||
BC093075.1 | - | 2 | 305 | 30402963 | ||
BP354016.1 | - | 2 | 430 | |||
CB962971.1 | - | 367 | ||||
CR456758.1 | - | 2 | 197 | |||
D14696.1 | - | 2 | 343 | |||
DR156048.1 | - | 2 | 738 |