Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286445.1 | NP_001273374.1 | 13 | 1889 | ||
NM_001286446.1 | NP_001273375.1 | 13 | 1992 | |||
NM_001286447.1 | NP_001273376.1 | 13 | 2089 | |||
NM_014722.3 | NP_055537.2 | 14 | 2177 | |||
NM_015864.3 | NP_056948.2 | 13 | 2089 | |||
XM_006715275.2 | XP_006715338.1 | 13 | 2106 | |||
XM_006715277.1 | XP_006715340.1 | 13 | 2027 | |||
XM_006715279.2 | XP_006715342.1 | 13 | 2084 | |||
XM_006715281.3 | XP_006715344.1 | 13 | 2106 | |||
XM_011515007.1 | XP_011513309.1 | 14 | 2615 | |||
XM_011515008.2 | XP_011513310.1 | 14 | 1946 | |||
XM_011515009.1 | XP_011513311.1 | 13 | 1818 | |||
XM_011515010.2 | XP_011513312.1 | 13 | 1859 | |||
XM_011515012.1 | XP_011513314.1 | 14 | 2256 | |||
XM_017011523.1 | XP_016867012.1 | 13 | 1781 | |||
XM_017011524.1 | XP_016867013.1 | 13 | 2035 | |||
XM_017011525.1 | XP_016867014.1 | 13 | 2027 | |||
XM_017011526.1 | XP_016867015.1 | 13 | 2027 | |||
XM_017011527.1 | XP_016867016.1 | 13 | 1859 | |||
GenBank mRNA | AB002384.1 | - | 14 | 1975 | ||
AK299047.1 | - | 13 | 1797 | |||
AK300138.1 | - | 13 | 2037 | |||
AK300950.1 | - | 13 | 1842 | |||
BC000930.2 | - | 3 | 753 | |||
BC001232.1 | - | 13 | 2079 | 3453890 | ||
U49187.1 | - | 13 | 1947 |