Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TBC1D5 SIRNA Assays ›
Gene Symbol:
TBC1D5
Gene Name:
TBC1 domain family member 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 17157162 - 17742739 on Build GRCh38
UniGene ID:
Hs.475629
Species:
Human
siRNA ID
HSS190616
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TBC1D5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134381.1
NP_001127853.1
11
889
NM_014744.2
NP_055559.1
10
2290
XM_005265611.1
XP_005265668.1
10
860
XM_005265612.1
XP_005265669.1
9
795
XM_005265614.1
XP_005265671.1
10
860
XM_005265615.1
XP_005265672.1
9
790
XM_005265616.4
XP_005265673.1
8
719
XM_006713430.1
XP_006713493.1
10
910
XM_011534281.1
XP_011532583.1
12
1171
XM_011534283.2
XP_011532585.1
11
949
XM_011534284.2
XP_011532586.1
11
994
XM_011534286.1
XP_011532588.1
10
926
XM_011534287.1
XP_011532589.1
10
908
XM_017007552.1
XP_016863041.1
13
1272
XM_017007553.1
XP_016863042.1
13
1302
XM_017007554.1
XP_016863043.1
18
1915
XM_017007555.1
XP_016863044.1
12
1154
XM_017007556.1
XP_016863045.1
11
1134
XM_017007557.1
XP_016863046.1
9
754
XM_017007558.1
XP_016863047.1
9
784
XM_017007559.1
XP_016863048.1
11
1134
XM_017007560.1
XP_016863049.1
11
964
XM_017007561.1
XP_016863050.1
9
795
XM_017007562.1
XP_016863051.1
10
908
XM_017007563.1
XP_016863052.1
8
714
XM_017007564.1
XP_016863053.1
9
790
XM_017007565.1
XP_016863054.1
10
1088
XM_017007566.1
XP_016863055.1
10
877
XM_017007567.1
XP_016863056.1
8
718
XM_017007568.1
XP_016863057.1
9
754
GenBank mRNA
AB449886.1
-
8
625
AK309374.1
-
6
550
AK310539.1
-
11
889
BC013145.1
-
9
802
4510019
D86965.1
-
10
2289
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of receptor internalization
intracellular protein transport
positive regulation of autophagy
regulation of vesicle fusion
retrograde transport, endosome to Golgi
response to starvation
activation of GTPase activity
mitophagy in response to mitochondrial depolarization
regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding
AP-2 adaptor complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0