Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTSS1 SIRNA Assays ›
Gene Symbol:
MTSS1
Gene Name:
metastasis suppressor 1
Gene Aliases:
MIM, MIMA, MIMB
Chromosome Location:
Chr.8: 124550770 - 124728507 on Build GRCh38
UniGene ID:
Hs.336994
Species:
Human
siRNA ID
HSS190620
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTSS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282971.1
NP_001269900.1
4, 5
855
NM_001282974.1
NP_001269903.1
4, 5
855
NM_014751.5
NP_055566.3
4, 5
855
XM_005251111.1
XP_005251168.1
4, 5
821
XM_005251113.1
XP_005251170.1
4, 5
821
XM_005251118.1
XP_005251175.1
4, 5
821
XM_006716700.1
XP_006716763.1
4, 5
821
XM_006716701.1
XP_006716764.1
4, 5
821
XM_006716702.1
XP_006716765.1
4, 5
821
XM_006716703.1
XP_006716766.1
4, 5
821
XM_006716704.1
XP_006716767.1
4, 5
821
XM_006716705.1
XP_006716768.1
4, 5
821
XM_006716706.1
XP_006716769.1
4, 5
821
XM_011517403.1
XP_011515705.1
4, 5
287
XM_011517404.2
XP_011515706.1
2, 3
11987
XM_017014086.1
XP_016869575.1
4, 5
821
XM_017014087.1
XP_016869576.1
4, 5
821
XM_017014088.1
XP_016869577.1
4, 5
821
XM_017014089.1
XP_016869578.1
4, 5
821
XM_017014090.1
XP_016869579.1
2, 3
1367
XM_017014091.1
XP_016869580.1
4, 5
255
XM_017014092.1
XP_016869581.1
4, 5
821
XM_017014093.1
XP_016869582.1
4, 5
821
XM_017014094.1
XP_016869583.1
4, 5
821
XM_017014095.1
XP_016869584.1
4, 5
821
XM_017014096.1
XP_016869585.1
4, 5
821
XM_017014097.1
XP_016869586.1
4, 5
821
XM_017014098.1
XP_016869587.1
4, 5
821
XM_017014099.1
XP_016869588.1
4, 5
821
XM_017014100.1
XP_016869589.1
4, 5
821
XM_017014101.1
XP_016869590.1
2, 3
480
GenBank mRNA
AB007889.2
-
1, 2
112
AF086645.1
-
4, 5
766
AK027015.1
-
4, 5
621
EF560715.1
-
4, 5
486
U50573.1
-
3, 4
240
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
movement of cell or subcellular component
plasma membrane organization
cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
microspike assembly
actin cytoskeleton organization
negative regulation of epithelial cell proliferation
renal tubule morphogenesis
cellular response to fluid shear stress
glomerulus morphogenesis
nephron tubule epithelial cell differentiation
xenophagy
epithelial cell proliferation involved in renal tubule morphogenesis
actin monomer binding
receptor binding
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0