Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270975.1 | NP_001257904.1 | 6 | 644 | ||
NM_001270976.1 | NP_001257905.1 | 7 | 759 | |||
NM_001270977.1 | NP_001257906.1 | 6 | 644 | |||
NM_001270978.1 | NP_001257907.1 | 3 | 272 | |||
NM_001270979.1 | NP_001257908.1 | 2 | 188 | |||
NM_014761.3 | NP_055576.2 | 6 | 644 | |||
GenBank mRNA | AB097052.1 | - | 6 | 595 | ||
AK057902.1 | - | 6 | 607 | |||
AK291388.1 | - | 6 | 629 | |||
AK293022.1 | - | 6 | 605 | |||
AK293040.1 | - | 7 | 747 | |||
AK295066.1 | - | 5 | 496 | |||
AK297088.1 | - | 2 | 580 | |||
AK297336.1 | - | 6 | 525 | |||
AK301918.1 | - | 2 | 150 | |||
BC000116.1 | - | 6 | 584 | 3350789 | ||
BC000430.2 | - | 6 | 594 | 2819828 | ||
BC004359.1 | - | 6 | 594 | 2819828 | ||
BC103745.1 | - | 6 | 622 | 6009274 | ||
BX364250.2 | - | 3 | 217 | |||
D79996.1 | - | 6 | 595 |