Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303264.1 | NP_001290193.1 | 3 | 3117 | ||
NM_014779.3 | NP_055594.1 | 4 | 3189 | |||
NR_130136.1 | 5 | 3346 | ||||
GenBank mRNA | AB014569.1 | - | 4 | 3189 | ||
AF201290.1 | - | 4 | 545 | |||
AF201291.1 | - | 3 | 244 | |||
AF201292.1 | - | 5 | 502 | |||
AK297383.1 | - | 4 | 1011 | |||
AK297748.1 | - | 3 | 446 | |||
BC044643.1 | - | 4 | 2523 | 5454441 |