Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286479.1 | NP_001273408.1 | 8 | 1041 | ||
NM_001286480.1 | NP_001273409.1 | 9 | 1163 | |||
NM_014783.4 | NP_055598.1 | 8 | 1782 | |||
NM_199357.1 | NP_955389.1 | 8 | 1782 | |||
GenBank mRNA | AK303025.1 | - | 9 | 1082 | ||
AK316437.1 | - | 8 | 1041 | |||
BC039563.2 | - | 8 | 1637 | 3913511 | ||
BC050545.1 | - | 8 | 1737 | |||
BC063444.1 | - | 8 | 1733 | |||
D87717.2 | - | 8 | 1782 |