Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009554.3 | NP_001009554.1 | 2 | 768 | ||
NM_001301647.1 | NP_001288576.1 | 2 | 843 | |||
NM_001301648.1 | NP_001288577.1 | 3 | 1030 | |||
NM_021647.7 | NP_067679.6 | 3 | 1274 | |||
XM_005263366.3 | XP_005263423.1 | 4 | 1208 | |||
XM_005263368.3 | XP_005263425.1 | 2 | 2382 | |||
XM_017008864.1 | XP_016864353.1 | 5 | 1152 | |||
XM_017008865.1 | XP_016864354.1 | 5 | 4268 | |||
XM_017008866.1 | XP_016864355.1 | 5 | 1342 | |||
XM_017008867.1 | XP_016864356.1 | 5 | 1148 | |||
XM_017008868.1 | XP_016864357.1 | 4 | 1423 | |||
XM_017008869.1 | XP_016864358.1 | 3 | 4255 | |||
XM_017008870.1 | XP_016864359.1 | 3 | 4215 | |||
XM_017008871.1 | XP_016864360.1 | 3 | 1155 | |||
GenBank mRNA | AB014526.1 | - | 3 | 1122 | ||
AF327560.1 | - | 2 | 754 | |||
AK290041.1 | - | 3 | 1184 | |||
AK308263.1 | - | 2 | 758 | |||
AY391838.1 | - | 2 | 1055 | |||
BC066912.1 | - | 2 | 737 | 5268480 |