Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014810.4 | NP_055625.4 | 21 | 4947 | ||
XM_011510191.2 | XP_011508493.1 | 21 | 4906 | |||
XM_011510198.2 | XP_011508500.1 | 21 | 4906 | |||
XM_017002929.1 | XP_016858418.1 | 21 | 4906 | |||
XM_017002930.1 | XP_016858419.1 | 21 | 4944 | |||
XM_017002931.1 | XP_016858420.1 | 21 | 4906 | |||
GenBank mRNA | AF287356.1 | - | 21 | 4731 | ||
AK000704.1 | - | 11 | 1865 | |||
BC048310.1 | - | 2 | 160 |