Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039479.1 | NP_001034568.1 | 4 | 578 | ||
XM_006720344.2 | XP_006720407.1 | 4 | 589 | |||
XM_011537415.2 | XP_011535717.1 | 5 | 651 | |||
XM_011537416.2 | XP_011535718.1 | 4 | 589 | |||
GenBank mRNA | AB002315.1 | - | 4 | 544 | ||
AK304217.1 | - | 4 | 555 | |||
BC032944.2 | - | 4 | 582 | 5267463 | ||
BI459956.1 | - | 4 | 587 |