Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286658.2 | NP_001273587.1 | 4 | 1627 | ||
NM_001286659.2 | NP_001273588.1 | 4 | 1627 | |||
NM_014832.4 | NP_055647.2 | 4 | 1627 | |||
XM_005266603.1 | XP_005266660.1 | 4 | 2105 | |||
XM_005266605.2 | XP_005266662.1 | 4 | 955 | |||
XM_006719903.3 | XP_006719966.1 | 4 | 1315 | |||
XM_011535331.1 | XP_011533633.1 | 4 | 2105 | |||
XM_017020882.1 | XP_016876371.1 | 4 | 836 | |||
XM_017020883.1 | XP_016876372.1 | 4 | 941 | |||
GenBank mRNA | AB011175.1 | - | 4 | 1573 | ||
AB449885.1 | - | 4 | 1226 | |||
AK307493.1 | - | 4 | 1627 | |||
BC151239.1 | - | 4 | 1565 | 8860512 | ||
FM207106.1 | - | 4 | 1226 | |||
FM207107.1 | - | 4 | 1226 |