Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278649.1 | NP_001265578.1 | 10 | 1157 | ||
NM_014835.3 | NP_055650.1 | 10 | 1176 | |||
NM_144498.2 | NP_653081.1 | 10 | 1212 | |||
XM_017028163.1 | XP_016883652.1 | 10 | 1212 | |||
XM_017028164.1 | XP_016883653.1 | 11 | 1517 | |||
XM_017028165.1 | XP_016883654.1 | 11 | 1517 | |||
XM_017028166.1 | XP_016883655.1 | 11 | 1266 | |||
XM_017028167.1 | XP_016883656.1 | 11 | 1280 | |||
XM_017028168.1 | XP_016883657.1 | 10 | 1212 | |||
XM_017028169.1 | XP_016883658.1 | 10 | 1146 | |||
XM_017028170.1 | XP_016883659.1 | 10 | 1146 | |||
GenBank mRNA | AB018315.1 | - | 10 | 1082 | ||
AF392447.1 | - | 10 | 1173 | |||
AK000230.1 | - | 10 | 1173 | |||
AK124174.1 | - | 4 | 533 | |||
AK291851.1 | - | 10 | 1140 | |||
AK296595.1 | - | 10 | 1118 | |||
AK315576.1 | - | 10 | 1173 | |||
AY028168.1 | - | 10 | 1082 | |||
BC000296.2 | - | 10 | 1116 | 2819652 | ||
BC004455.1 | - | 10 | 1116 | 2819652 | ||
BC018812.2 | - | 10 | 1116 | |||
CR456722.1 | - | 9 | 971 |