Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014840.2 | NP_055655.1 | 2 | 1710 | ||
GenBank mRNA | AB011109.1 | - | 2 | 1710 | ||
AK027302.1 | - | 3 | 571 | |||
AK289992.1 | - | 2 | 390 | |||
BC152462.1 | - | 2 | 1710 | 8862598 |