Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SV2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167580.1 | NP_001161052.1 | 10, 11 | 1670 | ||
NM_001323031.1 | NP_001309960.1 | 11, 12 | 2203 | |||
NM_001323032.1 | NP_001309961.1 | 11, 12 | 2512 | |||
NM_001323033.1 | NP_001309962.1 | 9, 10 | 2179 | |||
NM_001323036.1 | NP_001309965.1 | 10, 11 | 1489 | |||
NM_001323037.1 | NP_001309966.1 | 12, 13 | 2419 | |||
NM_001323038.1 | NP_001309967.1 | 11, 12 | 2331 | |||
NM_001323039.1 | NP_001309968.1 | 11, 12 | 2604 | |||
NM_001323040.1 | NP_001309969.1 | 11, 12 | 1850 | |||
NM_014848.5 | NP_055663.1 | 12, 13 | 2600 | |||
XM_005254998.3 | XP_005255055.1 | 11, 12 | 2265 | |||
XM_017022761.1 | XP_016878250.1 | 11, 12 | 2216 | |||
XM_017022762.1 | XP_016878251.1 | 11, 12 | 2351 | |||
GenBank mRNA | AB018278.1 | - | 10, 11 | 1873 | ||
AK294902.1 | - | 10, 11 | 1672 | |||
BC030011.2 | - | 12, 13 | 2237 | 4826206 | ||
CR457086.1 | - | 10, 11 | 1709 |