Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLRE1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271816.1 | NP_001258745.1 | 7 | 3306 | ||
NM_014881.4 | NP_055696.3 | 6 | 3624 | |||
XM_006718090.1 | XP_006718153.1 | 7 | 3318 | |||
XM_011540429.1 | XP_011538731.1 | 7 | 3374 | |||
GenBank mRNA | BC013124.2 | - | 6 | 3256 | 3048375 | |
BC062582.1 | - | 6 | 3256 | 5202821 | ||
D42045.1 | - | 6 | 3508 |