Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMMECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025580.1 | NP_001020751.1 | 4 | 811 | ||
NM_001171689.1 | NP_001165160.1 | 7 | 1109 | |||
NM_015365.2 | NP_056180.1 | 5 | 922 | |||
GenBank mRNA | AJ007014.1 | - | 5 | 1009 | ||
AK001703.1 | - | 5 | 445 | |||
AK091430.1 | - | 7 | 1109 | |||
AK314596.1 | - | 5 | 853 | |||
BC060813.1 | - | 4 | 744 | 30344824 | ||
BX648797.1 | - | 4 | 773 | |||
KU178496.1 | - | 4 | 730 | |||
KU178497.1 | - | 5 | 841 |