Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005114.2 | NP_005105.1 | 2 | 712 | ||
XM_011513913.2 | XP_011512215.1 | 2 | 5471 | |||
GenBank mRNA | AF019386.1 | - | 1 | 507 | ||
AK096823.1 | - | 2 | 1562 | |||
BC057803.1 | - | 2 | 694 | 5261530 |