Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204344.1 | NP_001191273.1 | 2 | 257 | ||
NM_005118.3 | NP_005109.2 | 4 | 550 | |||
GenBank mRNA | AF039390.1 | - | 1 | 1326 | ||
AF520785.1 | - | 4 | 556 | |||
AK291642.1 | - | 4 | 461 | |||
AK313721.1 | - | 4 | 525 | |||
AY434464.1 | - | 2 | 257 | |||
BC069435.1 | - | 4 | 505 | 7262219 | ||
BC074940.2 | - | 4 | 474 | 30915494 | ||
BC074941.2 | - | 4 | 474 | 30915271 | ||
BC104462.1 | - | 4 | 549 | 40030800 | ||
BC104463.1 | - | 4 | 550 | 40030803 |