Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004359.1 | NP_004350.1 | 1 | 301 | ||
XM_005259690.3 | XP_005259747.1 | 1 | 314 | |||
XM_006722952.2 | XP_006723015.1 | 1 | 314 | |||
GenBank mRNA | AK291554.1 | - | 1 | 303 | ||
BC009850.2 | - | 1 | 301 | 3942735 | ||
BC018143.2 | - | 1 | 304 | 3884179 | ||
BC023979.1 | - | 1 | 278 | 4103120 | ||
BT006659.1 | - | 1 | 102 | |||
L22005.1 | - | 1 | 288 |