Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000662.7 | NP_000653.3 | 3 | 428 | ||
NM_001160170.3 | NP_001153642.1 | 5 | 962 | |||
NM_001160171.3 | NP_001153643.1 | 4 | 812 | |||
NM_001160172.3 | NP_001153644.1 | 5 | 733 | |||
NM_001160173.3 | NP_001153645.1 | 4 | 578 | |||
NM_001160174.2 | NP_001153646.1 | 1 | 645 | |||
NM_001160175.3 | NP_001153647.1 | 5 | 696 | |||
NM_001160176.3 | NP_001153648.1 | 4 | 546 | |||
NM_001160179.2 | NP_001153651.1 | 5 | 632 | |||
NM_001291962.1 | NP_001278891.1 | 6 | 750 | |||
XM_006716410.3 | XP_006716473.1 | 3 | 1005 | |||
XM_011544687.1 | XP_011542989.1 | 6 | 1091 | |||
XM_011544688.1 | XP_011542990.1 | 5 | 941 | |||
XM_011544689.2 | XP_011542991.1 | 2 | 790 | |||
XM_017013947.1 | XP_016869436.1 | 8 | 1327 | |||
GenBank mRNA | AK290912.1 | - | 3 | 903 | ||
AV754344.1 | - | 3 | 421 | |||
BC013732.1 | - | 4 | 785 | |||
BC047666.1 | - | 3 | 407 | 6059458 | ||
BG655073.1 | - | 3 | 455 | |||
BX647521.1 | - | 4 | 521 | |||
D90041.1 | - | 1 | 411 |