Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145554.2 | NP_663529.2 | 7 | 740 | ||
GenBank mRNA | AK029008.1 | - | 7 | 705 | ||
AK148340.1 | - | 8 | 702 | |||
AK159800.1 | - | 7 | 684 | |||
AK167290.1 | - | 7 | 689 | |||
AK169675.1 | - | 2 | 173 | |||
BC021467.1 | - | 7 | 689 | 5067154 | ||
BC066808.1 | - | 7 | 683 | 30055031 | ||
BC067411.1 | - | 7 | 683 | 30092248 |