Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175305.4 | NP_780514.1 | 4 | 779 | ||
GenBank mRNA | AK033594.1 | - | 4 | 780 | ||
BC061228.1 | - | 5 | 819 | 30314668 |