Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmym6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285885.1 | NP_001272814.1 | 8 | 2202 | ||
NM_001285887.1 | NP_001272816.1 | 8 | 1119 | |||
NM_001285888.1 | NP_001272817.1 | 9 | 1402 | |||
NM_177462.5 | NP_803413.1 | 9 | 1395 | |||
GenBank mRNA | AK035122.1 | - | 1 | 267 | ||
AK035136.1 | - | 9 | 1397 | |||
AK081836.1 | - | 8 | 1109 | |||
AK082594.1 | - | 8 | 1926 | |||
AK170695.1 | - | 9 | 2418 | |||
BC034119.1 | - | 7 | 1047 | |||
BC040744.1 | - | 7 | 1047 | |||
BC094598.1 | - | 8 | 2175 | |||
BC140958.1 | - | 9 | 1270 | 9055570 | ||
BC144807.1 | - | 9 | 1277 |