Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpdl3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133888.2 | NP_598649.1 | 7 | 1042 | ||
GenBank mRNA | AK136602.1 | - | 6 | 704 | ||
AK154599.1 | - | 7 | 1068 | |||
AK166253.1 | - | 7 | 1018 | |||
BC009087.1 | - | 7 | 1042 | 2810972 |